Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 5, Clone: SP27, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21247305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21247305

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Reconstitution Protein A purified
Test Specificity Predicted to cross react with Bovine, Canine, Mouse, Porcine, Rat.
Regulatory Status RUO
Primary or Secondary Primary
Immunogen Synthetic peptide from C-terminus of human cytokeratin 5.
Purification Method Purified
Formulation TBS/1% BSA buffer pH 7.5
Clone SP27
Format Purified
Antigen Cytokeratin 5
Isotype IgG
Gene ID (Entrez) 3852