Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 5, Clone: SP27, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21247301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21247301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen Cytokeratin 5
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide from C-terminus of human cytokeratin 5.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3852
Test Specificity Predicted to cross react with Bovine, Canine, Mouse, Porcine, Rat.
Reconstitution Protein A purified
Clone SP27
Conjugate Unlabeled
Formulation TBS/1% BSA buffer pH 7.5