Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 18, Clone: R20-H, Novus Biologicals™

$307.00 - $307.00

Specifications

Antigen Cytokeratin 18
Clone R20-H
Immunogen Peptide derived from C-terminal region of human cytokeratin 18
Gene Accession No. P05783
Gene ID (Entrez) 3875
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17907210

 
novus biologicals canada
NBP179072100UL
100μL Each for $307.00
Description & Specifications

Specifications

Antigen Cytokeratin 18
Clone R20-H
Immunogen Peptide derived from C-terminal region of human cytokeratin 18
Gene Accession No. P05783
Gene ID (Entrez) 3875
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)