Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 18, Clone: R20-H, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179072200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907220

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Peptide derived from C-terminal region of human cytokeratin 18
Regulatory Status RUO
Clone R20-H
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P05783
Purification Method Affinity Purified
Quantity 200μL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3875
Antigen Cytokeratin 18
Host Species Rabbit