Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 18, Clone: R20-H, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179072100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907210

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen Peptide derived from C-terminal region of human cytokeratin 18
Storage Requirements Store at 4°C. Do not freeze.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Antigen Cytokeratin 18
Gene ID (Entrez) 3875
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 100μL
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Regulatory Status RUO
Purification Method Affinity Purified
Clone R20-H
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P05783
Host Species Rabbit