Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 17, Clone: V21-R, Novus Biologicals™

$309.05 - $397.00

Specifications

Antigen Cytokeratin 17
Clone V21-R
Immunogen Peptide derived from C-terminal region of human cytokeratin 17.
Gene Accession No. Q04695
Gene ID (Entrez) 3872
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17907110

 
novus biologicals canada
NBP179071100UL
100μL Each for $309.05

NBP17907120

 
novus biologicals canada
NBP179071200UL
200μL Each for $397.00
Description & Specifications

Specifications

Antigen Cytokeratin 17
Clone V21-R
Immunogen Peptide derived from C-terminal region of human cytokeratin 17.
Gene Accession No. Q04695
Gene ID (Entrez) 3872
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)