Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 17, Clone: V21-R, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179071100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907110

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 17
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Format Affinity Purified
Gene Accession No. Q04695
Immunogen Peptide derived from C-terminal region of human cytokeratin 17.
Isotype IgG
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3872
Clone V21-R
Cross Reactivity Human
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary or Secondary Primary