Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 17, Clone: V21-R, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179071200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907120

  • / Each
 

Description & Specifications

Specifications

Antigen Cytokeratin 17
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone V21-R
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. Q04695
Host Species Rabbit
Immunogen Peptide derived from C-terminal region of human cytokeratin 17.
Isotype IgG
Purification Method Affinity Purified
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3872