Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 16, Clone: R20-S, Novus Biologicals™

$309.05 - $309.05

Specifications

Antigen Cytokeratin 16
Clone R20-S
Immunogen Peptide derived from C-terminal region of human cytokeratin 16.
Gene Accession No. P08779
Gene ID (Entrez) 3868
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17907010

 
novus biologicals canada
NBP179070100UL
100μL Each for $309.05
Description & Specifications

Specifications

Antigen Cytokeratin 16
Clone R20-S
Immunogen Peptide derived from C-terminal region of human cytokeratin 16.
Gene Accession No. P08779
Gene ID (Entrez) 3868
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)