Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 16, Clone: R20-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179070100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907010

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 100μL
Host Species Rabbit
Cross Reactivity Human
Primary or Secondary Primary
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Antigen Cytokeratin 16
Regulatory Status RUO
Purification Method Affinity Purified
Immunogen Peptide derived from C-terminal region of human cytokeratin 16.
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3868
Clone R20-S
Gene Accession No. P08779