missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 14, Clone: D19-N, Novus Biologicals™

$309.05 - $309.05

Specifications

Antigen Cytokeratin 14
Clone D19-N
Immunogen Peptide derived from C-terminal region of human cytokeratin 14.
Gene ID (Entrez) 3861
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17906910 Novus Biologicals Canada
NBP179069100UL
100μL Each for $309.05
Specifications Description & Specifications

Specifications

Cytokeratin 14
Peptide derived from C-terminal region of human cytokeratin 14.
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary
Rabbit
Unlabeled
Affinity Purified
D19-N
3861
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Paraffin)