Please login to your online account to display your discounted pricing

Rabbit anti-Cytokeratin 14, Clone: D19-N, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179069500UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17906950

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 500μL
Isotype IgG
Primary or Secondary Primary
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone D19-N
Immunogen Peptide derived from C-terminal region of human cytokeratin 14.
Cross Reactivity Human
Antigen Cytokeratin 14
Gene ID (Entrez) 3861
Regulatory Status RUO
Purification Method Affinity Purified
Conjugate Unlabeled
Host Species Rabbit
Format Affinity Purified