Please login to your online account to display your discounted pricing

Rabbit anti-Cyclin D1, Clone: E16-I, Novus Biologicals™

$363.90 - $547.00

Specifications

Antigen Cyclin D1
Clone E16-I
Immunogen Peptide derived from C-terminal region of human cyclin D1.
Gene Accession No. P24385
Gene ID (Entrez) 595
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17901210

 
novus biologicals canada
NBP179012100UL
100μL Each for $363.90

NBP17901220

 
novus biologicals canada
NBP179012200UL
200μL Each for $547.00
Description & Specifications

Specifications

Antigen Cyclin D1
Clone E16-I
Immunogen Peptide derived from C-terminal region of human cyclin D1.
Gene Accession No. P24385
Gene ID (Entrez) 595
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)