Please login to your online account to display your discounted pricing

Rabbit anti-Cyclin D1, Clone: E16-I, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179012100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17901210

  • / Each
 

Description & Specifications

Specifications

Antigen Cyclin D1
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone E16-I
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Gene Accession No. P24385
Host Species Rabbit
Immunogen Peptide derived from C-terminal region of human cyclin D1.
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 595
Target Species Human