Please login to your online account to display your discounted pricing

Rabbit anti-Cyclin D1, Clone: E16-I, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179012200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17901220

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Format Affinity Purified
Gene Accession No. P24385
Host Species Rabbit
Immunogen Peptide derived from C-terminal region of human cyclin D1.
Quantity 200μL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone E16-I
Conjugate Unlabeled
Cross Reactivity Human
Antigen Cyclin D1
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 595