Please login to your online account to display your discounted pricing

Rabbit anti-CTRP7, Clone: K14-S, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13013201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13013201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Antigen CTRP7
Format Affinity Purified
Immunogen Peptide from near the carboxy terminus of human CTRP7.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Gene Accession No. Q9BXJ2
Host Species Rabbit
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 114905
Clone K14-S
Concentration 3.6mg/mL
Isotype IgG
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is specific to the carboxy terminus of CTRP7