Please login to your online account to display your discounted pricing

Rabbit anti-COX-2, Clone: SP21, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979610

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen COX-2
Format Supernatant
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of rat COX2.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Isotype IgG
Purification Method Supernatant
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 5743
Clone SP21