Please login to your online account to display your discounted pricing

Rabbit anti-COX-2, Clone: SP21, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979601

  • / Each
 

Description & Specifications

Specifications

Antigen COX-2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Primary or Secondary Primary
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Rabbit
Regulatory Status RUO
Isotype IgG
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Format Supernatant
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Supernatant
Clone SP21
Gene ID (Entrez) 5743
Monoclonal or Polyclonal Monoclonal
Immunogen Synthetic peptide corresponding to C-terminus of rat COX2.