missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Complement C4, Clone: A24-T, Novus Biologicals™

$332.66 - $332.66

Specifications

Antigen Complement C4
Clone A24-T
Immunogen Peptide derived from internal sequence of human C4.
Gene Accession No. P0C0L4
Gene ID (Entrez) 720
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17907510 Novus Biologicals Canada
NBP179075100UL
100μL Each for $332.66
Specifications Description & Specifications

Specifications

Complement C4
Peptide derived from internal sequence of human C4.
720
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
A24-T
P0C0L4
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary
Rabbit
Unlabeled
Affinity Purified