Please login to your online account to display your discounted pricing

Rabbit anti-Complement C4, Clone: A24-T, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179075200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907520

  • / Each
 

Description & Specifications

Specifications

Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgG
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Antigen Complement C4
Format Affinity Purified
Gene Accession No. P0C0L4
Host Species Rabbit
Immunogen Peptide derived from internal sequence of human C4.
Purification Method Affinity Purified
Quantity 200μL
Gene ID (Entrez) 720
Clone A24-T
Conjugate Unlabeled
Cross Reactivity Human
Monoclonal or Polyclonal Monoclonal