missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Complement C3d, Clone: E28-P, Novus Biologicals™

$332.66 - $332.66

Specifications

Antigen Complement C3d
Clone E28-P
Immunogen Peptide derived from N-terminal sequence of human C3.
Gene Accession No. P01024
Gene ID (Entrez) 718
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17907410 Novus Biologicals Canada
NBP179074100UL
100μL Each for $332.66
Specifications Description & Specifications

Specifications

Complement C3d
Peptide derived from N-terminal sequence of human C3.
718
Monoclonal
Store at 4°C. Do not freeze.
Unlabeled
Affinity Purified
E28-P
P01024
20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Primary
Rabbit
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)