Please login to your online account to display your discounted pricing

Rabbit anti-Complement C3d, Clone: E28-P, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179074200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17907420

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Antigen Complement C3d
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Host Species Rabbit
Immunogen Peptide derived from N-terminal sequence of human C3.
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P01024
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 718
Clone E28-P