Please login to your online account to display your discounted pricing

Rabbit anti-Claudin-1, Clone: SP128, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17451705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17451705

  • / Each
 

Description & Specifications

Specifications

Antigen Claudin-1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP128
Conjugate Unlabeled
Format Purified
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human claudin-1 protein.
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9076
Reconstitution Protein A purified
Target Species Human