Please login to your online account to display your discounted pricing

Rabbit anti-Chromogranin A, Clone: SP12, Novus Biologicals™

$274.00 - $332.66

Specifications

Antigen Chromogranin A
Clone SP12
Concentration 0.1mg/mL
Immunogen Recombinant protein encoding human chromogranin A.
Gene ID (Entrez) 1113
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21247001

 
novus biologicals canada
NBP21247001ML
0.1mL Each for $274.00

NBP21247005

 
novus biologicals canada
NBP21247005ML
0.5mL Each for $332.66
Description & Specifications

Specifications

Antigen Chromogranin A
Clone SP12
Concentration 0.1mg/mL
Immunogen Recombinant protein encoding human chromogranin A.
Gene ID (Entrez) 1113
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)