Please login to your online account to display your discounted pricing

Rabbit anti-Chromogranin A, Clone: SP12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21247005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21247005

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Chromogranin A
Format Supernatant
Immunogen Recombinant protein encoding human chromogranin A.
Isotype IgG
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Clone SP12
Conjugate Unlabeled
Cross Reactivity Human
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Host Species Rabbit
Purification Method Supernatant
Regulatory Status RUO
Test Specificity Predicted to cross react with Bovine, Porcine.
Concentration 0.1mg/mL