Please login to your online account to display your discounted pricing

Rabbit anti-CDK2, Clone: SP80, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen CDK2
Clone SP80
Immunogen Synthetic peptide corresponding to C-terminus of human CDK2 protein.
Gene Accession No. P24941
Gene ID (Entrez) 1017
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21248501

 
novus biologicals canada
NBP21248501ML
0.1mL Each for $309.05

NBP21248505

 
novus biologicals canada
NBP21248505ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen CDK2
Clone SP80
Immunogen Synthetic peptide corresponding to C-terminus of human CDK2 protein.
Gene Accession No. P24941
Gene ID (Entrez) 1017
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot