Please login to your online account to display your discounted pricing

Rabbit anti-CDK2, Clone: SP80, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248505

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human CDK2 protein.
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone SP80
Conjugate Unlabeled
Antigen CDK2
Formulation PBS/1% BSA buffer pH 7.6
Gene Accession No. P24941
Isotype IgG
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1017
Test Specificity Predicted to cross react with Bovine, Hamster, Mouse, Rat.
Reconstitution Protein A purified
Cross Reactivity Human