Please login to your online account to display your discounted pricing

Rabbit anti-CDK2, Clone: SP80, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248501

  • / Each
 

Description & Specifications

Specifications

Antigen CDK2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SP80
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Gene Accession No. P24941
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human CDK2 protein.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1017
Test Specificity Predicted to cross react with Bovine, Hamster, Mouse, Rat.
Reconstitution Protein A purified
Target Species Human