Please login to your online account to display your discounted pricing

Rabbit anti-CD99, Clone: SP119, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250805

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Formulation PBS/1% BSA buffer pH 7.6
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Recombinant full length human CD99 protein.
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4267
Reconstitution Protein A purified
Clone SP119
Antigen CD99