Please login to your online account to display your discounted pricing

Rabbit anti-CD99, Clone: SP119, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250801

  • / Each
 

Description & Specifications

Specifications

Antigen CD99
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP119
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Recombinant full length human CD99 protein.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4267
Reconstitution Protein A purified
Target Species Human