missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-CD8 alpha, Clone: P17-V, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen CD8 alpha
Clone P17-V
Immunogen Peptide derived from C-terminal region of human CD8
Gene Accession No. P01732
Gene ID (Entrez) 925
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17905510 Novus Biologicals Canada
NBP179055100UL
100μL Each for $363.90
Specifications Description & Specifications

Specifications

CD8 alpha
Peptide derived from C-terminal region of human CD8
925
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
P17-V
P01732
20mM Tris-HCl, pH 8.0 20mg/mL BSA.
Primary
Rabbit
Unlabeled
Affinity Purified