Please login to your online account to display your discounted pricing

Rabbit anti-CD7, Clone: C21-Q, Novus Biologicals™

$367.12 - $367.12

Specifications

Antigen CD7
Clone C21-Q
Immunogen Peptide derived from C-terminal region of human CD7.
Gene Accession No. P09564
Gene ID (Entrez) 924
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17908010

 
novus biologicals canada
NBP179080100UL
100μL Each for $367.12
Description & Specifications

Specifications

Antigen CD7
Clone C21-Q
Immunogen Peptide derived from C-terminal region of human CD7.
Gene Accession No. P09564
Gene ID (Entrez) 924
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)