Please login to your online account to display your discounted pricing

Rabbit anti-CD7, Clone: C21-Q, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179080200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17908020

  • / Each
 

Description & Specifications

Specifications

Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Immunogen Peptide derived from C-terminal region of human CD7.
Gene ID (Entrez) 924
Isotype IgG
Format Affinity Purified
Purification Method Affinity Purified
Regulatory Status RUO
Antigen CD7
Quantity 200μL
Cross Reactivity Human
Host Species Rabbit
Gene Accession No. P09564
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone C21-Q
Conjugate Unlabeled