Please login to your online account to display your discounted pricing

Rabbit anti-CD5, Clone: A25-G, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178994100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17899410

  • / Each
 

Description & Specifications

Specifications

Antigen CD5
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BS.
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Peptide derived from the C-terminal sequence of human CD10.
Primary or Secondary Primary
Gene ID (Entrez) 921
Clone A25-G
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal