Please login to your online account to display your discounted pricing

Rabbit anti-CD45, Clone: E19-G, Novus Biologicals™

$329.44 - $329.44

Specifications

Antigen CD45
Clone E19-G
Concentration 1.3mg/mL
Immunogen Peptide derived from C-terminal sequence of human CD45.
Gene Accession No. P08575
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13014501

 
novus biologicals canada
NBP13014501ML
0.1mL Each for $329.44
Description & Specifications

Specifications

Antigen CD45
Clone E19-G
Concentration 1.3mg/mL
Immunogen Peptide derived from C-terminal sequence of human CD45.
Gene Accession No. P08575
Gene ID (Entrez) 5788
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)