Please login to your online account to display your discounted pricing

Rabbit anti-CD45, Clone: E19-G, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13014501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13014501

  • / Each
 

Description & Specifications

Specifications

Antigen CD45
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone E19-G
Concentration 1.3mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P08575
Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human CD45.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity This product is specific to the C-terminus of CD45
Target Species Human