Please login to your online account to display your discounted pricing

Rabbit anti-CD3 epsilon, Clone: SP7, Novus Biologicals™

$309.05 - $769.59

Specifications

Antigen CD3 epsilon
Clone SP7
Immunogen A synthetic peptide corresponding tointernal region of the epsilon chain of human CD3 epsilon protein.
Gene ID (Entrez) 916
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14979401

 
novus biologicals canada
NBP14979401ML
0.1mL Each for $309.05

NBP14979410

 
novus biologicals canada
NBP14979410ML
1mL Each for $769.59
Description & Specifications

Specifications

Antigen CD3 epsilon
Clone SP7
Immunogen A synthetic peptide corresponding tointernal region of the epsilon chain of human CD3 epsilon protein.
Gene ID (Entrez) 916
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot