Please login to your online account to display your discounted pricing

Rabbit anti-CD3 epsilon, Clone: SP7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979410

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Immunogen A synthetic peptide corresponding tointernal region of the epsilon chain of human CD3 epsilon protein.
Gene ID (Entrez) 916
Test Specificity May react with Mouse.
Host Species Rabbit
Primary or Secondary Primary
Regulatory Status RUO
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Format Supernatant
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Antigen CD3 epsilon
Purification Method Supernatant
Isotype IgG
Clone SP7
Quantity 1mL