Please login to your online account to display your discounted pricing

Rabbit anti-CD3 epsilon, Clone: SP7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 916
Cross Reactivity Canine
Cross Reactivity Human
Isotype IgG
Clone SP7
Format Supernatant
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Immunogen A synthetic peptide corresponding tointernal region of the epsilon chain of human CD3 epsilon protein.
Regulatory Status RUO
Host Species Rabbit
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Supernatant
Antigen CD3 epsilon
Quantity 0.1mL
Test Specificity May react with Mouse.