Please login to your online account to display your discounted pricing

Rabbit anti-CD1a, Clone: L21-A, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen CD1a
Clone L21-A
Immunogen Peptide derived from C-terminal region
Gene Accession No. P06126
Gene ID (Entrez) 909
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17904310

 
novus biologicals canada
NBP179043100UL
100μL Each for $363.90
Description & Specifications

Specifications

Antigen CD1a
Clone L21-A
Immunogen Peptide derived from C-terminal region
Gene Accession No. P06126
Gene ID (Entrez) 909
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)