Please login to your online account to display your discounted pricing

Rabbit anti-CD1a, Clone: L21-A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179043200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17904320

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Format Affinity Purified
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen CD1a
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Gene Accession No. P06126
Immunogen Peptide derived from C-terminal region
Purification Method Affinity Purified
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 909
Clone L21-A
Conjugate Unlabeled