Please login to your online account to display your discounted pricing

Rabbit anti-CD1a, Clone: L21-A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179043100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17904310

  • / Each
 

Description & Specifications

Specifications

Antigen CD1a
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone L21-A
Conjugate Unlabeled
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA
Gene Accession No. P06126
Host Species Rabbit
Immunogen Peptide derived from C-terminal region
Isotype IgG
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Target Species Human