Please login to your online account to display your discounted pricing

Rabbit anti-CD19, Clone: SP110, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250405

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human CD19.
Clone SP110
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation PBS/1% BSA buffer pH 7.6
Gene ID (Entrez) 930
Purification Method Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Isotype IgG
Cross Reactivity Human
Quantity 0.5mL
Reconstitution Protein A purified
Format Purified
Regulatory Status RUO
Antigen CD19