Please login to your online account to display your discounted pricing

Rabbit anti-CD19, Clone: SP110, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250401

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Immunogen Synthetic peptide corresponding to C-terminus of human CD19.
Monoclonal or Polyclonal Monoclonal
Isotype IgG
Gene ID (Entrez) 930
Storage Requirements Store at 4°C. Do not freeze.
Clone SP110
Conjugate Unlabeled
Quantity 0.1mL
Formulation PBS/1% BSA buffer pH 7.6
Antigen CD19
Reconstitution Protein A purified
Purification Method Purified
Host Species Rabbit
Format Purified
Primary or Secondary Primary
Regulatory Status RUO