Please login to your online account to display your discounted pricing

Rabbit anti-CD163, Clone: SP96, Novus Biologicals™

$309.05 - $552.33

Specifications

Antigen CD163
Clone SP96
Immunogen Synthetic peptide corresponding to near N-terminus of human CD163 protein.
Gene ID (Entrez) 9332
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21249701

 
novus biologicals canada
NBP21249701ML
0.1mL Each for $309.05

NBP21249705

 
novus biologicals canada
NBP21249705ML
0.5mL Each for $552.33
Description & Specifications

Specifications

Antigen CD163
Clone SP96
Immunogen Synthetic peptide corresponding to near N-terminus of human CD163 protein.
Gene ID (Entrez) 9332
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)