Please login to your online account to display your discounted pricing

Rabbit anti-CD163, Clone: SP96, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249705

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Host Species Rabbit
Antigen CD163
Storage Requirements Store at 4°C. Do not freeze.
Format Purified
Immunogen Synthetic peptide corresponding to near N-terminus of human CD163 protein.
Primary or Secondary Primary
Gene ID (Entrez) 9332
Test Specificity Predicted to cross react with Bovine, Porcine.
Conjugate Unlabeled
Formulation PBS/1% BSA buffer pH 7.6
Quantity 0.5mL
Isotype IgG
Clone SP96
Reconstitution Protein A purified
Purification Method Purified