Please login to your online account to display your discounted pricing

Rabbit anti-CD163, Clone: SP96, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21249701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21249701

  • / Each
 

Description & Specifications

Specifications

Antigen CD163
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Immunogen Synthetic peptide corresponding to near N-terminus of human CD163 protein.
Primary or Secondary Primary
Test Specificity Predicted to cross react with Bovine, Porcine.
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9332
Reconstitution Protein A purified
Clone SP96
Isotype IgG