Please login to your online account to display your discounted pricing

Rabbit anti-CD117/c-kit, Clone: SP26, Novus Biologicals™

$309.05 - $630.24

Specifications

Antigen CD117/c-kit
Clone SP26
Immunogen Synthetic peptide corresponding to cytoplasmic domain of human CD117 protein
Gene ID (Entrez) 3815
Formulation Tissue culture Supernatant with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14979701

 
novus biologicals canada
NBP14979701ML
0.1mL Each for $309.05

NBP14979710

 
novus biologicals canada
NBP14979710ML
1mL Each for $630.24
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone SP26
Immunogen Synthetic peptide corresponding to cytoplasmic domain of human CD117 protein
Gene ID (Entrez) 3815
Formulation Tissue culture Supernatant with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)