Please login to your online account to display your discounted pricing

Rabbit anti-CD117/c-kit, Clone: SP26, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979710ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979710

  • / Each
 

Description & Specifications

Specifications

Formulation Tissue culture Supernatant with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Primary or Secondary Primary
Regulatory Status RUO
Quantity 1mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Antigen CD117/c-kit
Host Species Rabbit
Clone SP26
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Isotype IgG
Purification Method Affinity Purified
Immunogen Synthetic peptide corresponding to cytoplasmic domain of human CD117 protein
Gene ID (Entrez) 3815
Cross Reactivity Human