Please login to your online account to display your discounted pricing

Rabbit anti-CD117/c-kit, Clone: SP26, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979701

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP26
Primary or Secondary Primary
Gene ID (Entrez) 3815
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human
Antigen CD117/c-kit
Quantity 0.1mL
Isotype IgG
Immunogen Synthetic peptide corresponding to cytoplasmic domain of human CD117 protein
Formulation Tissue culture Supernatant with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Conjugate Unlabeled
Host Species Rabbit
Format Affinity Purified
Regulatory Status RUO
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal